(4-Bromo-2,6-dimethylphenoxy)acetonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3220-5G HC-3220-1G
COBBHC-3220-5GEA 0 GBP
COBBHC-3220-5G COBBHC-3220-1G
(4-Bromo-2,6-dimethylphenoxy)acetonitrile
(4-Bromo-2,6-dimethylphenoxy)acetonitrile
Formula: C₁₀H₁₀BrNO
MDL Number: MFCD09906423
CAS Number: 508189-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR