1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008381-5G 008381-1G
FLUO008381-5GEA 0 GBP
FLUO008381-5G FLUO008381-1G
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine
Formula: C₈H₈F₃N₃O₂
MDL Number: MFCD00052691
CAS Number: 120940-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR