Terephthalic acid dihydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018991-10G 018991-25G
FLUO018991-10GEA 0 GBP
FLUO018991-10G FLUO018991-25G
Terephthalic acid dihydrazide
Terephthalic acid dihydrazide
Formula: C₈H₁₀N₄O₂
MDL Number: MFCD00014762
CAS Number: 136-64-1
EINECS: 205-253-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR