Allylboronic acid pinacol ester, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane
M-Clarity™ quality level = MQ100
324647-10G
324647-1G
SIAL324647-10GEA
335
GBP
SIAL324647-10G
SIAL324647-1G
Allylboronic acid pinacol ester, Sigma-Aldrich®
Allylboronic acid pinacol ester
Formula:
C₉H₁₇BO₂ |
MDL Number:
MFCD00013347 CAS Number: 72824-04-5 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...