Allylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane
PN-8601-5G
PN-8601-1G
COBBPN-8601-5GEA
0
GBP
COBBPN-8601-5G
COBBPN-8601-1G
Allylboronic acid pinacol ester
Allylboronic acid pinacol ester
Formula:
C₉H₁₇BO₂ MW: 168.05 g/mol |
MDL Number:
MFCD00013347 CAS Number: 72824-04-5 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...