2-Amino-4,4,4-trifluorobutyric acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-2-Amino-4,4,4-trifluorobutyric acid, 2-Amino-4,4,4-trifluorobutanoic acid

001476-500MG 001476-1G
FLUO001476-500MGEA 0 GBP
FLUO001476-500MG FLUO001476-1G
2-Amino-4,4,4-trifluorobutyric acid
(±)-2-Amino-4,4,4-trifluorobutyric acid
Formula: C₄H₆F₃NO₂
MDL Number: MFCD00041417
CAS Number: 15959-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR