(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L08304.06 L08304.14
L08304.06EA 0 GBP
L08304.06 L08304.14
(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98%
(4-Ethoxybenzyl)triphenylphosphonium bromide
Formula: C₂₇H₂₆BrOP
MDL Number: MFCD00011834
CAS Number: 82105-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR