4-(Bromomethyl)tetrahydropyran

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AM-1015-5G AM-1015-1G
COBBAM-1015-5GEA 0 GBP
COBBAM-1015-5G COBBAM-1015-1G
4-(Bromomethyl)tetrahydropyran
4-(Bromomethyl)tetrahydropyran
Formula: C₆H₁₁BrO
MW: 179.0549 g/mol
MDL Number: MFCD03788561
CAS Number: 125552-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR