1-(Trifluoroacetyl)imidazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001329-5G 001329-25G
FLUO001329-5GEA 0 GBP
FLUO001329-5G FLUO001329-25G
1-(Trifluoroacetyl)imidazole
1-(Trifluoroacetyl)imidazole
Formula: C₅H₃F₃N₂O
MDL Number: MFCD00014501
CAS Number: 1546-79-8
EINECS: 216-282-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR