1-Methylimidazole-4-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Methyl-1H-imidazole-4-carboxylic acid
HI-1090-5G
HI-1090-1G
HI-1090-25G
COBBHI-1090-5GEA
0
GBP
COBBHI-1090-5G
COBBHI-1090-1G
COBBHI-1090-25G
1-Methylimidazole-4-carboxylic acid
1-Methylimidazole-4-carboxylic acid
Formula:
C₅H₆N₂O₂ MW: 126.11 g/mol |
MDL Number:
MFCD02179560 CAS Number: 41716-18-1 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...