1-Methylimidazole-2-carboxylic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-1H-imidazole-2-carboxylic acid

COBBHI-1093-5GEA 0 GBP
COBBHI-1093-5G COBBHI-1093-1G
1-Methylimidazole-2-carboxylic acid
1-Methylimidazole-2-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD01863421
CAS Number: 20485-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR