7-Amino-4-(trifluoromethyl)coumarin
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF571.5
004029-1G
004029-5G
CHMPCFF571.5EA
0
GBP
CHMPCFF571.5
FLUO004029-1G
FLUO004029-5G
7-Amino-4-(trifluoromethyl)coumarin
7-Amino-4-(trifluoromethyl)coumarin
Formula:
C₁₀H₆F₃NO₂ |
MDL Number:
MFCD00006858 CAS Number: 53518-15-3 EINECS: 258-599-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...