3-Phosphonopropionic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Phosphonopropanoic acid

003781-25G 003781-10G
FLUO003781-25GEA 0 GBP
FLUO003781-25G FLUO003781-10G
3-Phosphonopropionic acid
3-Phosphonopropionic acid
Formula: C₃H₇O₅P
MDL Number: MFCD00002770
CAS Number: 5962-42-5
EINECS: 227-738-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR