Diethyl (cyanomethyl)phosphonate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7521-5G OS-7521-100G OS-7521-25G
COBBOS-7521-5GEA 0 GBP
COBBOS-7521-5G COBBOS-7521-100G COBBOS-7521-25G
Diethyl (cyanomethyl)phosphonate
Diethyl (cyanomethyl)phosphonate
Formula: C₆H₁₂NO₃P
MW: 177.14 g/mol
MDL Number: MFCD00001893
CAS Number: 2537-48-6
EINECS: 219-806-0
UN: 3278
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR