Azidotrimethylsilane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Trimethylsilyl azide
M-Clarity™ quality level = MQ100
155071-10G
155071-50G
SIAL155071-10GEA
159
GBP
SIAL155071-10G
SIAL155071-50G
Azidotrimethylsilane, Sigma-Aldrich®
Azidotrimethylsilane
Formula:
C₃H₉N₃Si MW: 115.21 g/mol |
MDL Number:
MFCD00001986 CAS Number: 4648-54-8 EINECS: 225-078-5 UN: 1992 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...