3,3'-Bis(trifluoromethyl)benzhydrol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009042-1G
009042-5G
FLUO009042-1GEA
0
GBP
FLUO009042-1G
FLUO009042-5G
3,3'-Bis(trifluoromethyl)benzhydrol
3,3'-Bis(trifluoromethyl)benzhydrol
Formula:
C₁₅H₁₀F₆O |
MDL Number:
MFCD00039228 CAS Number: 1598-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...