3-(Trifluoromethyl)benzhydrol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007591-1G
007591-5G
FLUO007591-1GEA
0
GBP
FLUO007591-1G
FLUO007591-5G
3-(Trifluoromethyl)benzhydrol
3-(Trifluoromethyl)benzhydrol
Formula:
C₁₄H₁₁F₃O |
MDL Number:
MFCD00014397 CAS Number: 728-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...