5-Bromo-2'-deoxyuridine (Broxuridine)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

15240.01 15240.02
SERA15240.01EA 0 GBP
SERA15240.01 SERA15240.02
5-Bromo-2'-deoxyuridine (Broxuridine)
5-Bromo-2'-deoxyuridine (Broxuridine)
Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR