4-Chloro-2-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7945-1G PY-7945-5G
COBBPY-7945-1GEA 0 GBP
COBBPY-7945-1G COBBPY-7945-5G
4-Chloro-2-methoxypyridine
4-Chloro-2-methoxypyridine
Formula: C₆H₆ClNO
MDL Number: MFCD06738656
CAS Number: 72141-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR