4-Chloro-8-methylquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2762-5G HC-2762-1G
COBBHC-2762-5GEA 0 GBP
COBBHC-2762-5G COBBHC-2762-1G
4-Chloro-8-methylquinoline
4-Chloro-8-methylquinoline
Formula: C₁₀H₈ClN
MDL Number: MFCD00272330
CAS Number: 18436-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR