4,6-Dichloro-2-methylquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2148-1G
HC-2148-5G
COBBHC-2148-1GEA
0
GBP
COBBHC-2148-1G
COBBHC-2148-5G
4,6-Dichloro-2-methylquinoline
4,6-Dichloro-2-methylquinoline
Formula:
C₁₀H₇Cl₂N |
MDL Number:
MFCD00272372 CAS Number: 53342-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...