4,5-Dichloro-8-methylquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6027-5G HC-6027-1G
COBBHC-6027-5GEA 0 GBP
COBBHC-6027-5G COBBHC-6027-1G
4,5-Dichloro-8-methylquinoline
4,5-Dichloro-8-methylquinoline
Formula: C₁₀H₇Cl₂N
MDL Number: MFCD09787694
CAS Number: 948292-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR