2-Chloro-6-methylquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1305-5G PY-1305-1G PY-1305-25G
COBBPY-1305-5GEA 0 GBP
COBBPY-1305-5G COBBPY-1305-1G COBBPY-1305-25G
2-Chloro-6-methylquinoline
2-Chloro-6-methylquinoline
Formula: C₁₀H₈ClN
MW: 177.6302 g/mol
MDL Number: MFCD00462389
CAS Number: 4295-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR