(4-Methoxyphenoxy)acetonitrile ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
A10398.06
A10398.14
A10398.06EA
19.2
GBP
A10398.06
A10398.14
(4-Methoxyphenoxy)acetonitrile ≥97%
(4-Methoxyphenoxy)acetonitrile
Purity: 97%
CAS No.: 22446-12-4
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Refractive Index: 1.5270
Boiling Point: 106-110°/0.1mm
UN No.: 3276
MDL No.: MFCD00017528
Formula:
C₉H₉NO₂ Boiling Pt: 106…110 °C (0.1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00017528 CAS Number: 22446-12-4 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...