Perfluorobutane-1-sulphonyl fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007001-25G 007001-100G 007001-250G
FLUO007001-25GEA 0 GBP
FLUO007001-25G FLUO007001-100G FLUO007001-250G
Perfluorobutane-1-sulphonyl fluoride
Perfluorobutane-1-sulphonyl fluoride
Formula: C₄F₁₀O₂S
MDL Number: MFCD00007422
CAS Number: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR