1,1,1-Trifluoro-2-butanone 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

L16868.03 L16868.06
L16868.03EA 30.1 GBP
L16868.03 L16868.06
1,1,1-Trifluoro-2-butanone 96%
1,1,1-Trifluoro-2-butanone

Purity: 96% CAS No.: 381-88-4 Molecular Formula: C4H5F3O Formula Weight: 126.08 Flash Point: -17°(1°F) Refractive Index: 1.3070 Boiling Point: 44-45° Beilstein Reference: 1702220 UN No.: 1224 MDL No.: MFCD00190641

Formula: C₄H₅F₃O
MW: 126.08 g/mol
Boiling Pt: 44…45 °C
Density: 0.929
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190641
CAS Number: 381-88-4
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR