3-Pyridylacetonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Pyridineacetonitrile, 2-(Pyridin-3-yl)acetonitrile

M-Clarity™ quality level = MQ100

P66009-25G P66009-5G
SIALP66009-25GEA 102 GBP
SIALP66009-25G SIALP66009-5G
3-Pyridylacetonitrile, Sigma-Aldrich®
3-Pyridylacetonitrile
Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD00006406
CAS Number: 6443-85-2
EINECS: 229-241-1
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR