2-Pyridylacetonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Pyridineacetonitrile
246565-100G
246565-25G
246565-5G
SIAL246565-100GEA
356
GBP
SIAL246565-100G
SIAL246565-25G
SIAL246565-5G
2-Pyridylacetonitrile, Sigma-Aldrich®
2-Pyridylacetonitrile
Formula:
C₇H₆N₂ MW: 118.14 g/mol |
MDL Number:
MFCD00006346 CAS Number: 2739-97-1 EINECS: 220-364-6 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...