(2-Chlorophenoxy)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

022110-5G 022110-25G
FLUO022110-5GEA 0 GBP
FLUO022110-5G FLUO022110-25G
(2-Chlorophenoxy)acetonitrile
(2-Chlorophenoxy)acetonitrile
Formula: C₈H₆ClNO
MW: 167.60 g/mol
MDL Number: MFCD00017331
CAS Number: 43111-31-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR