1,2-Dibromoperfluoroheptane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010289-5G 010289-25G
FLUO010289-5GEA 0 GBP
FLUO010289-5G FLUO010289-25G
1,2-Dibromoperfluoroheptane
1,2-Dibromoperfluoroheptane
Formula: C₇Br₂F₁₄
MDL Number: MFCD03094409
CAS Number: 678-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR