2-Bromo-2-chloro-1,1,1-trifluoroethane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Bromo-1-chloro-2,2,2-trifluoroethane
M-Clarity™ quality level = MQ200
B4388-125ML
B4388-250ML
SIALB4388-125MLEA
218
GBP
SIALB4388-125ML
SIALB4388-250ML
2-Bromo-2-chloro-1,1,1-trifluoroethane, Sigma-Aldrich®
2-Bromo-2-chloro-1,1,1-trifluoroethane
Formula:
C₂HBrClF₃ |
MDL Number:
MFCD00009602 CAS Number: 151-67-7 EINECS: 205-796-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...