2-Aminoethyl dihydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
O-Phosphorylethanolamine
M-Clarity™ quality level = MQ200
P0503-100G
P0503-1G
P0503-25G
P0503-500G
P0503-5G
P0503-10MG
SIALP0503-100GEA
590
GBP
SIALP0503-100G
SIALP0503-1G
SIALP0503-25G
SIALP0503-500G
SIALP0503-5G
SIALP0503-10MG
2-Aminoethyl dihydrogen phosphate, Sigma-Aldrich®
2-Aminoethyl dihydrogen phosphate
Formula:
C₂H₈NO₄P MW: 141.0630 g/mol |
MDL Number:
MFCD00008178 CAS Number: 1071-23-4 EINECS: 213-988-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...