4-(Hydroxymethyl)indole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1H-Indol-4-yl)methanol

IN-0002-1G IN-0002-5G
COBBIN-0002-1GEA 0 GBP
COBBIN-0002-1G COBBIN-0002-5G
4-(Hydroxymethyl)indole
4-(Hydroxymethyl)indole
Formula: C₉H₉NO
MW: 147.1739 g/mol
MDL Number: MFCD01632220
CAS Number: 1074-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR