Succinic acid dihydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018990-10G 018990-25G
FLUO018990-10GEA 0 GBP
FLUO018990-10G FLUO018990-25G
Succinic acid dihydrazide
Succinic acid dihydrazide
Formula: C₄H₁₀N₄O₂
MDL Number: MFCD00007613
CAS Number: 4146-43-4
EINECS: 223-970-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR