Sebacic acid dihydrazide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018989-1G
018989-25G
FLUO018989-1GEA
0
GBP
FLUO018989-1G
FLUO018989-25G
Sebacic acid dihydrazide
Sebacic acid dihydrazide
Formula:
C₁₀H₂₂N₄O₂ |
MDL Number:
MFCD00039763 CAS Number: 925-83-7 EINECS: 213-126-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...