3,5-Difluorobenzyl mercaptan
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
024362-1G
024362-5G
FLUO024362-1GEA
0
GBP
FLUO024362-1G
FLUO024362-5G
3,5-Difluorobenzyl mercaptan
3,5-Difluorobenzyl mercaptan
Formula:
C₇H₆F₂S |
CAS Number:
358358-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...