(4-Fluorophenoxy)acetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF172.10
CFF172.50
010738-1G
010738-5G
CHMPCFF172.10EA
0
GBP
CHMPCFF172.10
CHMPCFF172.50
FLUO010738-1G
FLUO010738-5G
(4-Fluorophenoxy)acetonitrile
(4-Fluorophenoxy)acetonitrile
Formula:
C₈H₆FNO |
CAS Number:
24115-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...