3,5-Dibromo-2-methoxypyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

722731-5G 722731-1G
SIAL722731-5GEA 163 GBP
SIAL722731-5G SIAL722731-1G
3,5-Dibromo-2-methoxypyridine, Sigma-Aldrich®
3,5-Dibromo-2-methoxypyridine
Formula: C₆H₅Br₂NO
MW: 266.9200 g/mol
MDL Number: MFCD05663709
CAS Number: 13472-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR