4-(Trifluoromethoxy)toluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011226-5G 011226-1G
FLUO011226-5GEA 0 GBP
FLUO011226-5G FLUO011226-1G
4-(Trifluoromethoxy)toluene
4-(Trifluoromethoxy)toluene
Formula: C₈H₇F₃O
MDL Number: MFCD00042413
CAS Number: 706-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR