2-Chloro-5-nitropyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS1193G1 AS1193G5
ACTIAS1193G1EA 0 GBP
ACTIAS1193G1 ACTIAS1193G5
2-Chloro-5-nitropyrimidine
2-Chloro-5-nitropyrimidine
Formula: C₄H₂ClN₃O₂
MDL Number: MFCD04117995
CAS Number: 10320-42-0
EINECS: 233-703-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR