Magnesium acetate 1 mol/l in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

63052-100ML 63052-500ML
SIAL63052-100MLEA 131 GBP
SIAL63052-100ML SIAL63052-500ML
Magnesium acetate 1 mol/l in aqueous solution, Sigma-Aldrich®
Magnesium acetate
Formula: (H₃CCOO)₂Mg
MW: 142.39 g/mol
Melting Pt: 323
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00003539
CAS Number: 142-72-3
EINECS: 205-554-9
Merck Index: 12,05688

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR