3-(1,1,2,3,3,3-Hexafluoropropoxy)propene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Allyl 2H-hexafluoroprop-1-yl ether
007156-5G
007156-25G
FLUO007156-5GEA
0
GBP
FLUO007156-5G
FLUO007156-25G
3-(1,1,2,3,3,3-Hexafluoropropoxy)propene
3-(1,1,2,3,3,3-Hexafluoropropoxy)propene
Formula:
C₆H₆F₆O MW: 208.1 g/mol Boiling Pt: 86…89 °C (1013 hPa) Density: 1.305 g/cm³ (20 °C) |
MDL Number:
MFCD00043795 CAS Number: 59158-81-5 EINECS: 261-635-9 UN: 3271 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...