Flash chromatography columns, PuriFlash®, high capacity polymers
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF302H/800G PF302H/55G PF302H/40G PF302H/400G PF302H/2G PF302H/18G PF302H/150G PF302H/100G PF302H/10G
INTEPF302H/800GEA 10880 GBP
INTEPF302H/800G INTEPF302H/55G INTEPF302H/40G INTEPF302H/400G INTEPF302H/2G INTEPF302H/18G INTEPF302H/150G INTEPF302H/100G INTEPF302H/10G
Flash chromatography columns, PuriFlash®, high capacity polymers
Chromatography Columns
These flash chromatography columns, with high capacity polymers are designed to purify hydrophilic/hydrophobic polymer for polar and non polar compound <3 kDa.
  • Universal polymer with high surface area
  • High capacity reverse phase
  • Base material is SDVB polymer
  • pH stability ranges form 1 to 13
Order Now
SPECIFICATIONS
Environmentally Preferable