Flash chromatography columns, PuriFlash®, SCX bonded silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF50SCX/300G PF50SCX/6G PF50SCX/55G PF50SCX/35G PF50SCX/2500G PF50SCX/20G PF50SCX/175G PF50SCX/1250G PF50SCX/120G PF50SCX/450G
INTEPF50SCX/300GEA 1370 GBP
INTEPF50SCX/300G INTEPF50SCX/6G INTEPF50SCX/55G INTEPF50SCX/35G INTEPF50SCX/2500G INTEPF50SCX/20G INTEPF50SCX/175G INTEPF50SCX/1250G INTEPF50SCX/120G INTEPF50SCX/450G
Flash chromatography columns, PuriFlash®, SCX bonded silica
Chromatography Columns
These flash chromatography columns are suitable for the removal or isolation of basic compounds.SCX bonded silica has sulfonic acid functionality which retains weak basic compounds that have one or more positive charges.
  • Strong cation exchanger
  • Base material is high performance spherical silica
  • pH ranges form 1 to 7,5
Order Now
SPECIFICATIONS
Environmentally Preferable