Flash chromatography columns, PuriFlash®, high capacity, NH₂ silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF50NH2HCF0025 PF50NH2HC2500G PF50NH2HC1250G PF50NH2HC/6G PF50NH2HCF0040 PF50NH2HC/300G PF50NH2HC/450G PF50NH2HC/20G PF50NH2HC/175G PF50NH2HC/120G
INTEPF50NH2HCF0025EA 76 GBP
INTEPF50NH2HCF0025 INTEPF50NH2HC2500G INTEPF50NH2HC1250G INTEPF50NH2HC/6G INTEPF50NH2HCF0040 INTEPF50NH2HC/300G INTEPF50NH2HC/450G INTEPF50NH2HC/20G INTEPF50NH2HC/175G INTEPF50NH2HC/120G
Flash chromatography columns, PuriFlash®, high capacity, NH₂ silica
Chromatography Columns
These flash chromatography columns with high capacity NH₂ silica are used for polar amine compounds with reduced amine-silanol interaction and are also suitable for carbohydrates. They eliminate the use of TEA (triethylamine) for basic compounds.
  • High capacity for higher loading
  • Base material is high capacity spherical silica
  • pH stability ranges from 1,5 to 6,5