Flash chromatography columns, PuriFlash®, with silica, celite and C18
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PFDLSIHC08/4G PFDLSIHC08/40G PFDLSIHC08/25G PFDLSIHC08/12G PFDLSIHC0525G PFDLSIHC0512G PFDLSIHC05/4G PFDLSIHC05/40G PFDLIRC180840G PFDLIRC180825G PFDLIRC180812G PFDLIRC180525G PFDLIRC1808/4G PFDLIRC180512G PFDLIRC1805/4G PFDLE/4G PFDLE/40G PFDLE/25G PFDLE/12G PFDLCET08/4G PFDLCET08/40G PFDLCET08/25G PFDLIRC180540G PFDLCET08/12G
INTEPFDLSIHC08/4GEA 112 GBP
INTEPFDLSIHC08/4G INTEPFDLSIHC08/40G INTEPFDLSIHC08/25G INTEPFDLSIHC08/12G INTEPFDLSIHC0525G INTEPFDLSIHC0512G INTEPFDLSIHC05/4G INTEPFDLSIHC05/40G INTEPFDLIRC180840G INTEPFDLIRC180825G INTEPFDLIRC180812G INTEPFDLIRC180525G INTEPFDLIRC1808/4G INTEPFDLIRC180512G INTEPFDLIRC1805/4G INTEPFDLE/4G INTEPFDLE/40G INTEPFDLE/25G INTEPFDLE/12G INTEPFDLCET08/4G INTEPFDLCET08/40G INTEPFDLCET08/25G INTEPFDLIRC180540G INTEPFDLCET08/12G
Flash chromatography columns, PuriFlash®, with silica, celite and C18
Chromatography Columns
These dry load columns are simple to load and the luer fittings provide a universal fit for all flash systems.
  • Self contained, no adapters or special fittings
  • Hand tight to 15 bar/215 psi
  • Compatible with all flash systems
Delivery information: HC series is supplied with high capacity silica for maximum adsorption and celite series is supplied with celite and recommended for nonadsorptive loading.