1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, IPr.HCl

M-Clarity™ quality level = MQ200

574074-2G 574074-500MG
SIAL574074-2GEA 308 GBP
SIAL574074-2G SIAL574074-500MG
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, Sigma-Aldrich®
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride
Formula: C₂₇H₃₇ClN₂
MW: 425.05 g/mol
MDL Number: MFCD02684545
CAS Number: 250285-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR