1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, IPr.HCl
H27150.03
H27150.06
H27150.03EA
118
GBP
H27150.03
H27150.06
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥97%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride
Purity: 97%
CAS No.: 250285-32-6
Molecular Formula: C27H37ClN2
Formula Weight: 425.05
Melting Point: ca 280° dec.
MDL No.: MFCD02684545
Formula:
C₂₇H₃₇ClN₂ MW: 425.05 g/mol Melting Pt: ∼ 280 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02684545 CAS Number: 250285-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...