Triphenylvinylphosphonium bromide ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
L00708.06
L00708.14
L00708.22
L00708.06EA
37.6
GBP
L00708.06
L00708.14
L00708.22
Triphenylvinylphosphonium bromide ≥97%
Triphenylvinylphosphonium bromide
Purity: 97%
CAS No.: 5044-52-0
Molecular Formula: CH2=CHP(C5H5)3Br
Formula Weight: 369.25
Melting Point: 182-186°
Beilstein Reference: 4168860
Formula:
C₂₀H₁₈BrP Melting Pt: 182…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011807 CAS Number: 5044-52-0 EINECS: 225-740-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...