POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,3'-(Piperazine-1,4-diyl)bis(2-hydroxypropane-1-sulphonic acid), Piperazine-1,4-bis(2-hydroxypropanesulphonic acid) dihydrate, POPSO

6632.1 6632.2
ROTH6632.1EA 35.84 GBP
ROTH6632.1 ROTH6632.2
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid)
POPSO (β,β'-Dihydroxypiperazine-1,4-dipropanesulphonic acid)
Formula: C₁₀H₂₂N₂O₈S₂
MDL Number: MFCD00038350
CAS Number: 68189-43-5
EINECS: 269-199-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR